To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Form:Liquid
- Gene ID:UniprotID# P31327
- Antigen Synonyms:Carbamoyl-phosphate synthase [ammonia]|Carbamoylphosphate synthetase 1|Carbamoyl phosphate synthetase 1|Carbamoyl-phosphate synthetase I|Carbamoyl phosphate synthetase I|Carbamoyl phosphate synthase [ammonia]|Carbamoyl phosphate synthetase 1 mitochondrial|Carbamoylphosphate synthase|Carbamoyl phosphate synthase [ammonia] mitochondrial
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:~165 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human CPS1 protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77176-540
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [CPS1/8420] antibody to CPS1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: CPS1
Clonality: Monoclonal
Clone: CPS1/8420
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human