To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Form:Liquid
- Gene ID:UniprotID# P15085
- Antigen Synonyms:CBPA1_HUMAN|Procarboxypeptidase A1 pancreatic|carboxypeptidase A1 (pancreatic)|CPA|CPA1|Pancreatic carboxypeptidase A1|Carboxypeptidase A1|Carboxypeptidase A1 precursor
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:47 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human CPA1 protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77176-020
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [CPA1/8258R] antibody to Carboxypeptidase A for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Carboxypeptidase A
Clonality: Recombinant
Clone: CPA1/8258R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human