Anti-Carboxypeptidase A Mouse Recombinant Antibody [clone: rCPA1/8689]
Catalog # 77176-012
Supplier: ANTIBODIES.COM LLC
Specifications
- Form:Liquid
- Gene ID:UniprotID# P15085
- Antigen Synonyms:CBPA1_HUMAN|Procarboxypeptidase A1 pancreatic|carboxypeptidase A1 (pancreatic)|CPA|CPA1|Pancreatic carboxypeptidase A1|Carboxypeptidase A1|Carboxypeptidase A1 precursor
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:47 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human CPA1 protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77176-012
- Packaging:Plastic vial
Specifications
About this item
Recombinant mouse monoclonal [rCPA1/8689] antibody to Carboxypeptidase A for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Carboxypeptidase A
Clonality: Recombinant
Clone: rCPA1/8689
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human