Anti-BAP1 Rabbit Recombinant Antibody [clone: BAP1/8959R]
Catalog # 77175-884
Supplier: ANTIBODIES.COM LLC
Specifications
- Form:Liquid
- Gene ID:UniprotID# Q92560
- Antigen Synonyms:BRCA 1 associated protein 1|DKFZp686N04275|Cerebral protein 6|Cerebral protein 13|BRCA1 associated protein 1|BRCA1-associated protein 1|BAP1|BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase)|BAP1_HUMAN
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:91 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 630-729) of human BAP1 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77175-884
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [BAP1/8959R] antibody to BAP1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: BAP1
Clonality: Recombinant
Clone: BAP1/8959R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human