Anti-alpha B Crystallin Mouse Monoclonal Antibody [clone: CRYAB/4657]
Catalog # 77175-854
Supplier: ANTIBODIES.COM LLC
Specifications
- Form:Liquid
- Gene ID:UniprotID# P02511
- Antigen Synonyms:Crystallin alpha polypeptide 2|Alpha-crystallin B chain|Crystallin alpha B|CRYA2|Cryab|CRYAB_HUMAN|Alpha(B)-crystallin|Alpha B Crystallin|Alpha crystallin B chain
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:Predicted: 20 kDa/Observed: 22 - 30 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human full-length CRYAB protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77175-854
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [CRYAB/4657] antibody to alpha B Crystallin for WB and IHC-P with samples derived from Human.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: alpha B Crystallin
Clonality: Monoclonal
Clone: CRYAB/4657
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human