Anti-GFP Alpaca Polyclonal Antibody (Magnetic Beads)
Catalog # 77209-882
Supplier: Avantor
Specifications
- Cat. No.:77209-882
Specifications
About this item
Alpaca polyclonal antibody to GFP (Magnetic Beads) for IP, ChIP, RIP and Co-IP with samples derived from Species Independent.
- Validated Applications: IP, ChIP, RIP, Co-IP
- Recommended Dilutions: Binding Capacity: 0.4 mg GFP protein/ml, CoIP: 500 µl (20 reactions), RIP: 500 µl (20 reactions), ChIP: 500 µl (20 reactions)
Type: Primary
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: Magnetic Beads
Epitope:
Host: Alpaca
Isotype: VHH
Reactivity: Species Independent