Anti-alpha Adaptin Rabbit Monoclonal Antibody [clone: ARC54447]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC54447] antibody to alpha Adaptin for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:2,000 to 1:20,000
Type: Primary
Antigen: alpha Adaptin
Clonality: Monoclonal
Clone: ARC54447
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Related Information
Frequently Bought Together