Anti-Lamin A Rabbit Monoclonal Antibody [clone: ARC5001-08]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC5001-08] antibody to Lamin A for WB, IHC, ICC/IF and IP with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ICC/IF, IP
- Recommended Dilutions: WB: 1:10,000 to 1:60,000, IHC: 1:50 to 1:400, ICC/IF: 1:50 to 1:200, IP: 1:10,000 to 1:1:120,000
Type: Primary
Antigen: Lamin A
Clonality: Monoclonal
Clone: ARC5001-08
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Frequently Bought Together