Anti-JunD Rabbit Monoclonal Antibody [clone: ARC1408]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC1408] antibody to JunD for WB, IP and ChIP with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IP, ChIP
- Recommended Dilutions: WB: 1:500 to 1:2,000, IP: 1:500 to 1:1,000, ChIP: 1:500 to 1:1,000
Type: Primary
Antigen: JunD
Clonality: Monoclonal
Clone: ARC1408
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Frequently Bought Together