Anti-CCDC98 Rabbit Monoclonal Antibody [clone: ARC2760]
: Avantor
Rabbit monoclonal [ARC2760] antibody to CCDC98 for WB and IP with samples derived from Human.
- Validated Applications: WB, IP
- Recommended Dilutions: WB: 1:100 to 1:500, IP: 1:100 to 1:500
Type: Primary
Antigen: CCDC98
Clonality: Monoclonal
Clone: ARC2760
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together