Anti-PAK4 + PAK5 + PAK6 (phospho Ser474 + Ser560 + Ser602) Rabbit Monoclonal Antibody [clone: ARC1596]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC1596] antibody to PAK4 + PAK5 + PAK6 (phospho Ser474 + Ser560 + Ser602) for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: PAK4 + PAK5 + PAK6
Clonality: Monoclonal
Clone: ARC1596
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Related Information
Frequently Bought Together