Anti-Cyclin A1 + Cyclin A2 Rabbit Monoclonal Antibody [clone: ARC2642]
Rabbit monoclonal [ARC2642] antibody to Cyclin A1 + Cyclin A2 for WB and IP with samples derived from Human and Mouse.
- Validated Applications: WB, IP
- Recommended Dilutions: WB: 1:500 to 1:1,000, IP: 1:500 to 1:1,000
Type: Primary
Antigen: Cyclin A1 + Cyclin A2
Clonality: Monoclonal
Clone: ARC2642
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Frequently Bought Together