Anti-SLC14A1/UTE Rabbit Monoclonal Antibody [clone: ARC60284]
Supplier: Avantor
Certificates
About this item
Rabbit monoclonal [ARC60284] antibody to SLC14A1/UTE for WB, IHC and ICC/IF with samples derived from Human and Mouse.
- Validated Applications: WB, IHC, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: SLC14A1/UTE
Clonality: Monoclonal
Clone: ARC60284
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Related Information
Frequently Bought Together