Anti-beta II Tubulin Rabbit Monoclonal Antibody [clone: ARC0227]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC0227] antibody to beta II Tubulin for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: beta II Tubulin
Clonality: Monoclonal
Clone: ARC0227
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Related Information
Frequently Bought Together