Anti-Cdc6 (phospho Ser54) Rabbit Monoclonal Antibody [clone: ARC1693]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC1693] antibody to Cdc6 (phospho Ser54) for WB with samples derived from Human and Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: Cdc6
Clonality: Monoclonal
Clone: ARC1693
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Related Information
Frequently Bought Together