Anti-ZBTB7A Rabbit Monoclonal Antibody [clone: ARC2320]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC2320] antibody to ZBTB7A for WB, IHC and ICC/IF with samples derived from Human.
- Validated Applications: WB, IHC, ICC/IF
- Recommended Dilutions: WB: 1:1,000 to 1:3,000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: ZBTB7A
Clonality: Monoclonal
Clone: ARC2320
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77228-580
- Antigen Symbol:
- ZBTB7A
- Antigen Name:
- DKFZp547O146
- Antigen Synonyms:
- Leukemia/lymphoma-related factor|HIV-1 inducer of short transcripts-binding factor 1|Factor binding IST protein 1|FBI-1|Factor that binds to inducer of short transcripts protein 1|LRF|HIV-1 1st-binding protein 1|FBI1|HIV-1 inducer of short transcripts binding protein
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC2320
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- UniprotID# O95365
- Isotype:
- IgG
- Western Blot:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence within amino acids 450-550 of human ZBTB7A/FBI-1/LRF (O95365)
- Sequence:
- NYDLKNHMRVHTGLRPYQCDSCCKTFVRSDHLHRHLKKDGCNGVPSRRGRKPRVRGGAPDPSPGATATPGAPAQPSSPDARRNGQEKHFKDEDEDEDVASP
- Form:
- Liquid
- Molecular Weight:
- 75 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
- Tested Applications:
- IHC, ICC/IF
Specifications
Frequently Bought Together