Anti-TSPAN6 Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to TSPAN6 for WB with samples derived from Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: TSPAN6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77228-552
- Antigen Symbol:
- TSPAN6
- Antigen Name:
- 6720473L21Rik
- Antigen Synonyms:
- DKFZp469J2433|A15 homolog|MGC117923|T245|Putative NF-kappa-B-activating protein 321|Putative NF kappa B activating protein 321|AI316786|T245 protein|MGC133784
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# O43657
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 120-200 of human TSPAN6 (NP_003261.1)
- Sequence:
- NSFKNNYEKALKQYNSTGDYRSHAVDKIQNTLHCCGVTDYRDWTDTNYYSEKGFPKSCCKLEDCTPQRDADKVNNEGCFIK
- Form:
- Liquid
- Molecular Weight:
- 28 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together