Anti-hnRNP D/AUF1 Rabbit Monoclonal Antibody [clone: ARC59955]
Supplier: Avantor
Certificates
About this item
Rabbit monoclonal [ARC59955] antibody to hnRNP D/AUF1 for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ICC/IF
- Recommended Dilutions: WB: 1:1,000 to 1:5,000, IHC: 1:500 to 1:1,000, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: hnRNP D/AUF1
Clonality: Monoclonal
Clone: ARC59955
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Related Information
Frequently Bought Together