Anti-PAK3 Rabbit Monoclonal Antibody [clone: ARC1955]
Rabbit monoclonal [ARC1955] antibody to PAK3 for WB and IHC with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC
- Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200
Type: Primary
Antigen: PAK3
Clonality: Monoclonal
Clone: ARC1955
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Frequently Bought Together