Anti-c-Myc Rabbit Monoclonal Antibody [clone: ARC0412]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC0412] antibody to c-Myc for WB, IP and ChIP with samples derived from Human and Mouse.
- Validated Applications: WB, IP, ChIP
- Recommended Dilutions: WB: 1:500 to 1:1,000, ChIP: 1:50 to 1:200, CUT&Tag: 16 cells/2µg
Type: Primary
Antigen: c-Myc
Clonality: Monoclonal
Clone: ARC0412
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Frequently Bought Together