Anti-PTGR2 Goat Polyclonal Antibody
77076-290
- Antibody Type:Primary
- Antigen Name:15 oxoprostaglandin 13 reductase
- Antigen Symbol:PTGR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Pig
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:DKFZp686P10120|FLJ99229|PGR2|PRG-2|15-oxoprostaglandin 13-reductase|Prostaglandin reductase 2|FLJ39091|15 oxoprostaglandin delta 13 reductase|PGR 2
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-PDNINEGQVQVRT
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human PTGR2 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77076-290
- Packaging:Plastic vial
Goat polyclonal antibody to PTGR2 for ELISA and WB with samples derived from Porcine.
- Validated Applications: ELISA, WB
Type: Primary
Antigen: PTGR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Porcine