Anti-ERLIN1 Goat Polyclonal Antibody
77072-778
- Antibody Type:Primary
- Antigen Name:Band 7 23-211 Keo4 (Interim) similar to C.elegans protein
- Antigen Symbol:ERLIN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:C10orf69|Endoplasmic reticulum lipid raft-associated protein 1|ERLIN1|chromosome 10 open reading frame 69|Erlin-1|Erlin 1|KE04|ERLN1_HUMAN|ER lipid raft associated 1
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-EALEPSGENVIQNKE
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human ERLIN1 (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77072-778
- Packaging:Plastic vial
Goat polyclonal antibody to ERLIN1 for ELISA and WB with samples derived from Human.
- Validated Applications: ELISA, WB
Type: Primary
Antigen: ERLIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human