Anti-GPR83 Goat Polyclonal Antibody
Catalog # 77064-688
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:G protein coupled receptor 72
- Antigen Symbol:GPR83
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:GPR83_HUMAN|G protein coupled receptor 83|GPR83|Glucocorticoid induced receptor|G-protein coupled receptor 72|GIR|GPR 72|GPR72|GPR 83
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-RRYGAESQNPTVK
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human GPR83 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77064-688
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to GPR83 for ELISA and WB with samples derived from Human, Mouse and Rat.
- Validated Applications: ELISA, WB
Type: Primary
Antigen: GPR83
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat