Anti-TOM1L2 Goat Polyclonal Antibody
Catalog # 77065-106
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:FLJ32746
- Antigen Symbol:TOM1L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:TOM1|TM1L2_HUMAN|TOM1-like 2|TOM1L 2|Target of myb1-like 2 (chicken)|TOM1-like protein 2|Target of myb1 like 2|Target of Myb-like protein 2|Srebf1
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:CRKKPERSEDALFAL
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human TOM1L2 (C terminal)
- Tested Applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77065-106
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to TOM1L2 for ELISA, WB and IHC with samples derived from Human.
- Validated Applications: ELISA, WB, IHC
Type: Primary
Antigen: TOM1L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human