Anti-TCP1 Goat Polyclonal Antibody
Catalog # 77079-850
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:AI528772
- Antigen Symbol:TCP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:c-cpn|Ccta|D6S230E|MGC133746|CCT alpha|CCT-alpha|CCT1|CCT|CCTalpha
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-SYEDAVHSGALND
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human TCP1 (C terminal)
- Tested Applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77079-850
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to TCP1 for ELISA, WB, IHC, IF and Flow Cytometry with samples derived from Human.
- Validated Applications: ELISA, WB, IHC, IF, Flow Cytometry
Type: Primary
Antigen: TCP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human