Anti-WRNIP1 Goat Polyclonal Antibody
77076-514
- Antibody Type:Primary
- Antigen Name:ATPase WRNIP 1
- Antigen Symbol:WRNIP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:FLJ22526|Werner helicasae interacting protein|RP11 420G6.2|bA420G6.2|Werner helicase-interacting protein 1|Werner helicasae interacting protein 1|Putative helicase RUVBL|Werner helicase interacting protein isoform 2|ATPase WRNIP1
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:EELRGVDFFKQRRC
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human WRNIP1 (C terminal)
- Tested Applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77076-514
- Packaging:Plastic vial
Goat polyclonal antibody to WRNIP1 for ELISA, WB and IHC with samples derived from Human.
- Validated Applications: ELISA, WB, IHC
Type: Primary
Antigen: WRNIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human