Anti-ARPC4 Goat Polyclonal Antibody
77067-914
- Antibody Type:Primary
- Antigen Name:Actin related protein 2/3 complex subunit 4
- Antigen Symbol:ARPC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Actin related protein 2/3 complex, 20-KD subunit|ARPC4|ARC20|20kDa|20 kD|ARPC 4|Arp2/3 complex 20 kDa subunit|Actin-related protein 2/3 complex subunit 4|Arp2/3 protein complex subunit p20
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-ERHNKPEVEVR
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human ARPC4 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77067-914
- Packaging:Plastic vial
Goat polyclonal antibody to ARPC4 for ELISA and WB with samples derived from Human.
- Validated Applications: ELISA, WB
Type: Primary
Antigen: ARPC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human