Anti-SIM2 Goat Polyclonal Antibody
77071-462
- Antibody Type:Primary
- Antigen Name:bHLHe15
- Antigen Symbol:SIM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:SIM|SIM 2|Single minded homolog 2 (Drosophila)|MGC119447|SIM2|Class E basic helix loop helix protein 15|SIM2_HUMAN|Class E basic helix-loop-helix protein 15|Single minded homolog 2
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-SDLLYTPSYS
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human SIM2 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77071-462
- Packaging:Plastic vial
Goat polyclonal antibody to SIM2 for ELISA and WB with samples derived from Mouse.
- Validated Applications: ELISA, WB
Type: Primary
Antigen: SIM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse