Anti-GRIPAP1 Goat Polyclonal Antibody
Catalog # 77064-830
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:DKFZp434P0630
- Antigen Symbol:GRIPAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:GRASP-1|GRASP1|GRAP1_HUMAN|GRIP associated protein 1|GRIP 1 associated protein 1|GRIP1-associated protein 1|GRIP1 associated protein 1|GRASP 1|DXImx47e
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:LREQTTGLAAELQ-C
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human GRIPAP1 (N terminal)
- Tested Applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77064-830
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to GRIPAP1 for ELISA and IHC with samples derived from Human.
- Validated Applications: ELISA, IHC
Type: Primary
Antigen: GRIPAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human