Anti-KDM2B Goat Polyclonal Antibody
77073-492
- Antibody Type:Primary
- Antigen Name:CXXC-type zinc finger protein 2
- Antigen Symbol:KDM2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:F-box protein FBL10|F box and leucine rich repeat protein 10|F box protein FBL10|JEMMA (Jumonji domain EMSY interactor methyltransferase motif) protein|JHDM1B|F-box and leucine-rich repeat protein 10|CXXC2|F-box/LRR-repeat protein 10|FBXL10
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-FGQVEEKLLQKLS
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human KDM2B (C terminal)
- Tested Applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77073-492
- Packaging:Plastic vial
Goat polyclonal antibody to KDM2B for ELISA and IHC with samples derived from Human.
- Validated Applications: ELISA, IHC
Type: Primary
Antigen: KDM2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human