Anti-CPNE1 Goat Polyclonal Antibody
77063-880
- Antibody Type:Primary
- Antigen Name:Copine 1
- Antigen Symbol:CPNE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:CPN1|CPN 1|CPNE 1|CopineI|COPN1|Copine-1|Copine1|COPN 1|Copine I
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-APLKPLPPSAKDPA
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human CPNE1 (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77063-880
- Packaging:Plastic vial
Goat polyclonal antibody to CPNE1 for ELISA and WB with samples derived from Human.
- Validated Applications: ELISA, WB
Type: Primary
Antigen: CPNE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human