Anti-STK38 Goat Polyclonal Antibody
Catalog # 77063-938
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:NDR
- Antigen Symbol:STK38
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:1 protein kinase|Nuclear Dbf2-related protein|Ser/Thr kinase-like protein|Serine/threonine protein kinase 38|Nuclear Dbf2-related kinase 1|1|nuclear Dbf2-related 1|Nuclear Dbf2 related kinase 1
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-KPTVATSNHPET
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human STK38 (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77063-938
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to STK38 for ELISA and WB with samples derived from Human and Rat.
- Validated Applications: ELISA, WB
Type: Primary
Antigen: STK38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat