Anti-Cytochrome P450 1A1 Goat Polyclonal Antibody
Catalog # 77065-376
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:AHH
- Antigen Symbol:Cytochrome P450 1A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:AHRR|CYP1|CYPIA1|CP1A1_HUMAN|cyp1a1|Aryl hydrocarbon hydroxylase|CYP 1|Cytochrome P1 450 dioxin inducible|CP11
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-QDRKLDENANVQLSD
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Mouse cyp1a1 (internal region)
- Tested Applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77065-376
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to Cytochrome P450 1A1 for ELISA, WB, IF and IHC with samples derived from Human, Mouse and Rat.
- Validated Applications: ELISA, WB, IF, IHC
Type: Primary
Antigen: Cytochrome P450 1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat