Anti-NKX3-2 Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to NKX3-2 for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2000
Type: Primary
Antigen: NKX3-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 76860-656
- Antigen Symbol:
- NKX3-2
- Antigen Name:
- Bagpipe homeobox 1
- Antigen Synonyms:
- MGC138171|BAPX1|BAPX 1|Bagpipe homeobox homolog 1|Homeobox protein NK-3 homolog B|Homeobox protein Nkx-3.2|Bagpipe homeobox protein homolog 1|Homeobox protein Nkx 3.2|Bagpipe homeobox, Drosophila, homolog of, 1
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- UniprotID# P78367
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 1 - 180 of human NKX3 - 2 (NP_001180.1)
- Sequence:
- MAVRGANTLTSFSIQAILNKKEERGGLAAPEGRPAPGGTAASVAAAPAVCCWRLFGERDAGALGGAEDSLLASPAGTRTAAGRTAESPEGWDSDSALSEENESRRRCADARGASGAGLAGGSLSLGQPVCELAASKDLEEEAAGRSDSEMSASVSGDRSPRTEDDGVGPRGAHVSALCSG
- Molecular Weight:
- 35 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 50% Glycerol
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together