Anti-Amphiregulin Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to Amphiregulin for WB and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:1000, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: Amphiregulin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 76856-024
- Antigen Symbol:
- Amphiregulin
- Antigen Name:
- A REG
- Antigen Synonyms:
- A REG|AR|AREG_HUMAN|Amphiregulin|Colorectum cell derived growth factor|Amphiregulin B|AREGB|Colorectum cell-derived growth factor|CRDGF|AREG
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# P15514
- Isotype:
- IgG
- Western Blot:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 20 - 252 of human Amphiregulin (NP_001648.1)
- Sequence:
- SGHYAAGLDLNDTYSGKREPFSGDHSADGFEVTSRSEMSSGSEISPVSEMPSSSEPSSGADYDYSEEYDNEPQIPGYIVDDSVRVEQVVKPPQNKTESENTSDKPKRKKKGGKNGKNRRNRKKKNPCNAEFQNFCIHGECKYIEHLEAVTCKCQQEYFGERCGEKSMKTHSMIDSSLSKIALAAIAAFMSAVILTAVAVITVQLRRQYVRKYEGEAEERKKLRQENGNVHAIA
- Form:
- Liquid
- Molecular Weight:
- 43 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
- Tested Applications:
- ICC/IF
Specifications
Frequently Bought Together