Anti-CD89 Rabbit Polyclonal Antibody
Catalog # 76858-098
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Immunoglobulin alpha Fc receptor
- Antigen Symbol:CD89
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:UniprotID# P24071
- Antigen Synonyms:Fc fragment of IgA, receptor for|FCAR_HUMAN|Fc alpha receptor|CD89|IgA Fc receptor|FCAR
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Molecular Weight:40 kDa
- Sequence:LMIIKNSTYREIGRRLKFWNETDPEFVIDHMDANKAGRYQCQYRIGHYRFRYSDTLELVVTGLYGKPFLSADRGLVLMPGENISLTCSSAHIPFDRFSLAKEGELSLPQHQSGEHPANFSLGPVDLNVSGIYRCYGWYNRSPYLWSFPSNALELVVTDSIHQDYTTQNLIRMAVAGLVLVALLAILVENWHSHTALNKEASADVAEPSWSQQMCQPGLTFARTPSVCK
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 60 - 287 of human FCAR (NP_001991.1)
- Purification:Affinity purification
- Cat. No.:76858-098
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to CD89 for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:200 to 1:1000
Type: Primary
Antigen: CD89
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human