Anti-NUP98 Mouse Monoclonal Antibody [clone: 13C2 + 21A10]
Catalog # 77080-970
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:96 kDa nucleoporin
- Antigen Symbol:NUP98
- Clonality:Monoclonal
- Clone:13C2 + 21A10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:S. cerevisiae,S. pombe,Tetrahymena,Human
- Western Blot:Yes
- Size:100 µg
- Epitope:FGxxN for clone 13C2 and GLF for clone 21A10
- Form:Liquid
- Gene ID:UniprotID# P52948
- Antigen Synonyms:Nucleoporin Nup96|nucleoporin 98kDa|ADIR2|Nucleoporin 98kD|ADAR2|Nuclear pore complex protein Nup98 Nup96|GLFG-repeat containing nucleoporin|98 kDa nucleoporin|Nuclear pore complex protein Nup96
- Storage Buffer:Supplied in Phosphate Buffered Saline with 50% Glycerol (filter sterilised, without Sodium Azide, and carrier free)
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptides containing conserved N-terminal sequence, GLFG, of Nup98 protein of Tetrahymena thermophila.
Peptide 1: 1-MFGNTGGGGLFGNTQTQQTGGGLFGQPQQ-29
Peptide 2: 646-SNPTQGGGLFGAANPGLGG-664.
- Tested Applications:ICC/IF
- Purification:Affinity purification
- Cat. No.:77080-970
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal (13C2 + 21A10) antibody to NUP98 for WB and ICC/IF with samples derived from Human, Saccharomyces cerevisiae, Schizosaccharomyces pombe and Tetrahymena.
- Validated Applications: WB, ICC/IF
Type: Primary
Antigen: NUP98
Clonality: Monoclonal
Clone: 13C2 + 21A10
Conjugation: Unconjugated
Epitope: FGxxN for clone 13C2 and GLF for clone 21A10.
Host: Mouse
Isotype: IgG1
Reactivity: Human, Saccharomyces cerevisiae, Schizosaccharomyces pombe, Tetrahymena