Anti-HP1 alpha Rabbit Polyclonal Antibody
Catalog # 77076-774
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Antigen p25
- Antigen Symbol:HP1 alpha
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:UniprotID# P45973
- Antigen Synonyms:CBX5|Epididymis luminal protein 25|CBX5_HUMAN|CG8409|HEL25|Chromobox homolog 5|Chromobox 5|Chromobox protein homolog 5|Chromobox homolog 5 (HP1 alpha homolog, Drosophila)
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol (filter sterilised, without Sodium Azide, and carrier free)
- Sequence:CEDAENKEKETAKS
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:0.37 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to human HP1 alpha protein (a.a. 179-191)
- Purification:Affinity purification
- Cat. No.:77076-774
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to HP1 alpha for WB and IF with samples derived from Human.
- Validated Applications: WB, IF
- Recommended Dilutions: WB: 1:1000 to 1:3000, IF: 1:200 to 1:1000
Type: Primary
Antigen: HP1 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human