Anti-beta Galactosidase Rabbit Polyclonal Antibody
77073-060
- Antibody Type:Primary
- Antigen Name:Acid beta galactosidase
- Antigen Symbol:beta Galactosidase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:E. coli
- Western Blot:Yes
- Size:200 µg
- Form:Liquid
- Gene ID:UniprotID# B7UJI9
- Antigen Synonyms:EBP|Beta D galactosidase|GLB1|JW0335|ELNR1|ECK0341|Galactosidase beta 1|Beta gal|Beta galactosidase 1
- Storage Buffer:Supplied in Phosphate Buffered Saline with 50% Glycerol
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:2 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Full length E. coli beta Galactosidase
- Tested Applications:ICC/IF
- Purification:Protein A chromatography
- Cat. No.:77073-060
- Packaging:Plastic vial
Rabbit polyclonal antibody to beta Galactosidase for WB, IP, ICC/IF and ELISA with samples derived from Escherichia coli beta Galactosidase and beta Gal Tagged proteins..
- Validated Applications: WB, IP, ICC/IF, ELISA
- Recommended Dilutions: WB: 1:1000 to 1:2000, IP: 1:200 to 1:500, ICC/IF: 1:200 to 1:500, ELISA: 1:2000 to 1:3000
Type: Primary
Antigen: beta Galactosidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli beta Galactosidase and beta Gal Tagged proteins.