Anti-GST Rabbit Polyclonal Antibody
Catalog # 77067-044
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Glutathione S Transferase
- Antigen Symbol:GST
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen Synonyms:Glutathione S-transferase class-mu 26 kDa isozyme|GST epsilon|A1|GST class alpha 2|GST|GST class alpha 1|GST gamma|A2|GSTA1
- Storage Buffer:Supplied as antiserum with 0.05% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full length Glutathione S transferase protein (a.a. 1-212)
- Cat. No.:77067-044
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to GST for WB, IP and ELISA.
- Validated Applications: WB, IP, ELISA
- Recommended Dilutions: WB: 1:2000 to 1:10000
Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: