Anti-5-Methylcytosine Mouse Monoclonal Antibody [clone: 5MC-CD]
Catalog # 77064-190
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:5-methylcytosine
- Antigen Symbol:5-Methylcytosine
- Clonality:Monoclonal
- Clone:5MC-CD
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM
- Reactivity:Multiple
- Size:100 µg
- Form:Liquid
- Storage Buffer:Supplied in Phosphate Buffered Saline with 50% Glycerol
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:5-Methylcytosine conjugated to bovine serum albumin
- Tested Applications:ICC/IF, Southern Blot
- Purification:Affinity purification
- Cat. No.:77064-190
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal (5MC-CD) antibody to 5-Methylcytosine for ICC/IF, Southern Blot and FC with samples derived from DNA with 5-Methylcytosine (methylated DNA) and any species..
- Validated Applications: ICC/IF, Southern Blot, FC
- Recommended Dilutions: ICC/IF: 1:50 to 1:100
Type: Primary
Antigen: 5-Methylcytosine
Clonality: Monoclonal
Clone: 5MC-CD
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: DNA with 5-Methylcytosine (methylated DNA), any species.