Anti-IgG3 Rat Monoclonal Antibody [clone: KT94] (HRP (Horseradish Peroxidase))
Catalog # 77041-492
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:DKFZp686H11213
- Antigen Symbol:IgG3
- Clonality:Monoclonal
- Clone:KT94
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P03987
- Antigen Synonyms:FLJ40834|Heavy chain disease protein|FLJ40036|FLJ40587|HDC|Ig gamma 3 chain C region|FLJ39988|FLJ40789|FLJ40253
- Storage Buffer:Supplied in Phosphate Buffered Saline with 0.01% Thimerosal
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Mouse IgG3
- Purification:IgG is purified through a Protein G column and labelled with horseradish peroxidase
- Cat. No.:77041-492
- Packaging:Plastic vial
Specifications
About this item
Rat monoclonal (KT94) antibody to IgG3 (HRP) for ELISA and WB with samples derived from Mouse.
- Validated Applications: ELISA, WB
- Recommended Dilutions: ELISA: 1/1000, WB: 1/500 to 1/5000
Type: Primary
Antigen: IgG3
Clonality: Monoclonal
Clone: KT94
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse