Anti-RAB6B Rat Monoclonal Antibody [clone: KT79]
Catalog # 77041-450
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:9630015C03
- Antigen Symbol:RAB6B
- Clonality:Monoclonal
- Clone:KT79
- Conjugation:Unconjugated
- Host:Rat
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P61294
- Antigen Synonyms:D9Bwg0185e|OTTHUMP00000216372|RAB6B|Ras-related protein Rab-6B|Ras related protein Rab 6B|RAB6B_HUMAN|RAB6B member RAS oncogene family|C330006L04Rik|AI844641
- Storage Buffer:Supplied in Phosphate Buffered Saline with 0.09% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant mouse RAB6B
- Purification:Antibody is purified from culture supernatant using a protein G column
- Cat. No.:77041-450
- Packaging:Plastic vial
Specifications
About this item
Rat monoclonal (KT79) antibody to RAB6B for WB and IF with samples derived from Mouse.
- Validated Applications: WB, IF
- Recommended Dilutions: WB: 1/500 to 1/10000, IF: 1/100 to 1/1000
Type: Primary
Antigen: RAB6B
Clonality: Monoclonal
Clone: KT79
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse