Anti-MAPRE3 Rat Monoclonal Antibody [clone: KT53]
77041-720
- Antibody Type:Primary
- Antigen Name:APC binding protein
- Antigen Symbol:MAPRE3
- Clonality:Monoclonal
- Clone:KT53
- Conjugation:Unconjugated
- Host:Rat
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9UPY8
- Antigen Synonyms:MAPRE 3|EBF3 S|End-binding protein 3|EBF3|EBF 3|End binding protein 3|EB1 protein family member 3|EB3|EB 3
- Storage Buffer:Supplied in Phosphate Buffered Saline with 0.09% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Expressed human EB3 fragment
- Purification:Antibody is purified from culture supernatant using a protein G column
- Cat. No.:77041-720
- Packaging:Plastic vial
Rat monoclonal (KT53) antibody to MAPRE3 for WB and IP with samples derived from Human.
- Validated Applications: WB, IP
- Recommended Dilutions: WB: 1/500 to 1/5000, IP: 2 µg/ml
Type: Primary
Antigen: MAPRE3
Clonality: Monoclonal
Clone: KT53
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human