Anti-SARS-CoV-2 Spike Glycoprotein Human Recombinant Antibody [clone: CR3022]
77188-130
- Antibody Type:Primary
- Antigen Name:SARS-CoV-2 Spike Glycoprotein
- Antigen Symbol:SARS-CoV-2 Spike Glycoprotein
- Clonality:Recombinant
- Clone:CR3022
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Human
- Isotype:IgG1
- Reactivity:SARS-CoV-2
- Size:100 µg
- Form:Lyophilized
- Gene ID:UniprotID# P0DTC2
- Storage Buffer:Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% trehalose is added as a protectant before lyophilization
- Storage Temperature:Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Concentration:Reconstitution dependent
- Shipping Temperature:Shipped on blue ice at 4 °C
- Cat. No.:77188-130
- Packaging:Plastic vial
Recombinant human monoclonal [CR3022] antibody to SARS-CoV-2 Spike Glycoprotein for ELISA and flow cytometry with samples derived from SARS-CoV-2.
- Validated applications: ELISA, Flow cytometry
- Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100
Type: Primary
Antigen: SARS-CoV-2 Spike Glycoprotein
Clonality: Recombinant
Clone: CR3022
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1
Reactivity: SARS-CoV-2