Anti-Mesothelin Recombinant Recombinant Antibody
Catalog # 77188-114
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CAK 1
- Antigen Symbol:Mesothelin
- Clonality:Recombinant
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Recombinant
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:Lyophilized
- Gene ID:UniprotID# Q13421
- Antigen Synonyms:MPF|CAK1|Megakaryocyte potentiating factor|Msln|MSLN_HUMAN|cleaved form|CAK1 antigen|Mesothelin|Mesothelin isoform 1 precursor
- Storage Buffer:Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% trehalose is added as a protectant before lyophilization
- Storage Temperature:Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Concentration:Reconstitution dependent
- Shipping Temperature:Shipped on blue ice at 4 °C
- Cat. No.:77188-114
- Packaging:Plastic vial
Specifications
About this item
Recombinant chimeric monoclonal [Amatuximab Biosimilar] antibody to Mesothelin for ELISA and flow cytometry with samples derived from human.
- Validated applications: ELISA, Flow cytometry
- Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100
Type: Primary
Antigen: Mesothelin
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human