Anti-ErbB2/HER2 Recombinant Recombinant Antibody
Catalog # 77188-062
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ErbB2/HER2
- Antigen Symbol:ErbB2/HER2
- Clonality:Recombinant
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Recombinant
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:Lyophilized
- Gene ID:UniprotID# P04626
- Storage Buffer:Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% trehalose is added as a protectant before lyophilization
- Storage Temperature:Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Concentration:Reconstitution dependent
- Shipping Temperature:Shipped on blue ice at 4 °C
- Cat. No.:77188-062
- Packaging:Plastic vial
Specifications
About this item
Recombinant humanized monoclonal [Trastuzumab Biosimilar] antibody to ErbB2/HER2 for ELISA and flow cytometry with samples derived from human.
- Validated applications: ELISA, Flow cytometry
- Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100
Type: Primary
Antigen: ErbB2/HER2
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human