Anti-TSLP Human Recombinant Antibody
Catalog # 77187-954
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Thymic stromal lymphopoietin
- Antigen Symbol:TSLP
- Clonality:Recombinant
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Human
- Isotype:IgG2
- Reactivity:Human
- Size:100 µg
- Form:Lyophilized
- Gene ID:UniprotID# Q969D9
- Antigen Synonyms:TSLP protein|TSLP_HUMAN|Thymic stromal lymphopoietin protein TSLP
- Storage Buffer:Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% trehalose is added as a protectant before lyophilization
- Storage Temperature:Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Concentration:Reconstitution dependent
- Shipping Temperature:Shipped on blue ice at 4 °C
- Cat. No.:77187-954
- Packaging:Plastic vial
Specifications
About this item
Recombinant human monoclonal [Tezepelumab Biosimilar] antibody to TSLP for ELISA and flow cytometry with samples derived from human.
- Validated applications: ELISA, Flow cytometry
- Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100
Type: Primary
Antigen: TSLP
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG2
Reactivity: Human