To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:ILT-4
- Antigen Symbol:ILT-4
- Clonality:Recombinant Monoclonal
- Clone:MK-4830
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Chimera
- Isotype:IgG4
- Reactivity:Human
- Size:100 µg
- Environmentally Preferable:
- Form:Lyophilized
- Gene ID:UniprotID# Q8N423
- Storage Buffer:Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% trehalose is added as a protectant before lyophilization
- Storage Temperature:Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Concentration:Reconstitution dependent
- Shipping Temperature:Shipped on blue ice at 4 °C
- Cat. No.:77187-902
- Packaging:Plastic vial
Specifications
About this item
Recombinant humanized monoclonal [MK-4830] antibody to ILT-4 for ELISA and flow cytometry with samples derived from human.
- Validated applications: ELISA, Flow cytometry
- Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100
Type: Primary
Antigen: ILT-4
Clonality: Recombinant
Clone: MK-4830
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG4
Reactivity: Human