Anti-CD166 Chimera Recombinant Monoclonal Antibody
Catalog # 77187-850
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Activated leukocyte cell adhesion molecule
- Antigen Symbol:CD166
- Clonality:Recombinant Monoclonal
- Conjugation:Unconjugated
- Host:Chimera
- Isotype:IgG1
- Reactivity:Human
- Form:Lyophilized
- Antigen Synonyms:ALCAM|ALCAM protein|CD 166|CD166|CD166 antigen|CD166_HUMAN|FLJ3851|FLJ38514|MEMD
- Storage Buffer:Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% trehalose is added as a protectant before lyophilization
- Storage Temperature:Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Cat. No.:77187-850
Specifications
About this item
Recombinant humanized monoclonal [Praluzatamab Biosimilar] antibody to CD166 for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1:100
Type: Primary
Antigen: CD166
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human